Order Entry
Slovenia
ContactUsLinkComponent
Levetiracetam Hydroxybutyramide Analog, Pharmaceutical Analytical Impurities
Levetiracetam Hydroxybutyramide Analog, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates
Levetiracetam Hydroxybutyramide Analog, Pharmaceutical Analytical Impurities
Supplier:  USP

Specifications

  • CAS No.:
    1868118-63-1
  • Synonym(s):
    (S)-N-(1-amino-1-oxobutan-2-yl)-4-hydroxybutanamide
  • Description:
    S-Levetiracetam Hydroxybutyramide Analog

Specifications

Frequently Bought Together