Order Entry
Slovenia
ContactUsLinkComponent
Phentolamine Related Compounds, Reference Standard
Phentolamine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Phentolamine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • Synonym(s):
    N-(2-Aminoethyl)-2-[(3-hydroxyphenyl)(4-tolyl)amino]acetamide
  • Description:
    Phentolamine Related Compound A
  • CAS No.:
    65-28-1

Specifications

Frequently Bought Together