Order Entry
Slovenia
ContactUsLinkComponent
 
Phenobarbital Related Compounds, Reference Standard
 :  USP
 
Phenobarbital Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    69125-70-8
  • Synonym(s):
    5-Ethyl-2,6-diimino-5-phenyltetrahydropyrimidin-4(1H)-one
  • Description:
    Phenobarbital Related Compound A

 

Frequently Bought Together