Order Entry
Slovenia
ContactUsLinkComponent
Celecoxib, Pharmaceutical Analytical Impurities
Celecoxib, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates
Celecoxib, Pharmaceutical Analytical Impurities
Supplier:  USP

Specifications

  • CAS No.:
    170569-99-0
  • Synonym(s):
    4-(5-(o-tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulfonamide
  • Description:
    o-Celecoxib

Specifications

Frequently Bought Together