Nitrophenylchlorothiophene Carboxylate, Pharmaceutical Analytical Impurities
Catalog # 1A01590.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1450877-56-1
- Description:Nitrophenylchlorothiophene Carboxylate
- Synonym(s):4-nitrophenyl 5-chlorothiophene-2-carboxylate
- Description:Nitrophenylchlorothiophene Carboxylate
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.