N-Isopropyl Carvedilol, Pharmaceutical Analytical Impurities
1A00230.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1246819-01-1
- Description:N-Isopropyl Carvedilol
- Synonym(s):1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl](1-methylethyl)amino]-2-propanol
- Description:N-Isopropyl Carvedilol
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.