Order Entry
Slovenia
ContactUsLinkComponent
Nefazodone Related Compounds, Reference Standard
Nefazodone Related Compounds, Reference Standard
Supplier:  USP
Certificates
Nefazodone Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • Synonym(s):
    2-[3-[4-(4-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-3H-1,2,4-triazol-3-one hydrochloride
  • Description:
    Nefazodone Related Compound B
  • CAS No.:
    57059-62-8

Specifications

Frequently Bought Together