N6-(1-Iminoethyl)-D-lysine, Pharmaceutical Analytical Impurities
Catalog # 1A00530.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1844123-52-9
- Description:N6-(1-Iminoethyl)-D-lysine
- Synonym(s):(2R)-2-[(2S)-2-Aminobutanamido]butanamide
- Description:N6-(1-Iminoethyl)-D-lysine
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.