Order Entry
Slovenia
ContactUsLinkComponent
Loxapine Related Compounds, Reference Standard
Loxapine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Loxapine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    27833-64-3
  • UN No.:
    2811
  • Description:
    Loxapine Succinate
  • Synonym(s):
    3-Chloro-11-(4-methylpiperazin-1-yl)dibenzo[b,f][1,4]oxazepine

Specifications

Frequently Bought Together