Order Entry
Slovenia
ContactUsLinkComponent
Dicloxacillin Related Compounds, Reference Standard
Dicloxacillin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Dicloxacillin Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    3919-76-4
  • Synonym(s):
    3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
  • Description:
    Dicloxacillin Related Compound D

Specifications

Frequently Bought Together