Bis(2-methylaminoethyl) Ether Hydrochloride, Pharmaceutical Analytical Impurities
1A01330.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:295796-12-2
- Description:Bis(2-methylaminoethyl) Ether Hydrochloride
- Synonym(s):2,2'-oxybis(N-methylethan-1-amine), hydrochloride
- Description:Bis(2-methylaminoethyl) Ether Hydrochloride
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.