Order Entry
Slovenia
ContactUsLinkComponent
 
Aminophenylmorpholinone, Pharmaceutical Analytical Impurities
  1A01530.
 :  USP
 :  
Aminophenylmorpholinone, Pharmaceutical Analytical Impurities
  1A01530.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    438056-69-0
  • Description:
    Aminophenylmorpholinone
  • Synonym(s):
    4-(4-aminophenyl)morpholin-3-one
  • Description:
    Aminophenylmorpholinone

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.