Order Entry
Slovenia
ContactUsLinkComponent
Acetylcholine Chloride, Reference Standard
Acetylcholine Chloride, Reference Standard
Catalog # 1008501.
Supplier:  USP
CAS Number:  
Acetylcholine Chloride, Reference Standard
Catalog # 1008501.
Supplier:  USP
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • CAS No.:
    60-31-1
  • Description:
    Acetylcholine Chloride
  • Formula:
    C₇H₁₆NO₂Cl
  • Description:
    Acetylcholine Chloride

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard