SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene) ≥90%, Sigma-Aldrich®
Catalog # SIAL803472-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:360.49 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
CAS Number:112241-19-7
Formula:C₁₈H₂₀N₂O₂S₂
MDL Number:MFCD00083161