Anti-GOD Goat Polyclonal Antibody
Catalog # NOVUNBP2-44186
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glucose Oxidase
- Antigen symbol:GOD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Fungus
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:GOD|Beta D Glucose Oxygen 1 Oxido Reductase|Glucose Oxyhydrase
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Glucose Oxidase [Aspergillus niger]
- Purification:IgG purified
- Pk:100 µG
Specifications
About this item
The glucose oxidase antibody from Novus Biologicals is a goat polyclonal antibody to glucose oxidase. This antibody reacts with fungi. The glucose oxidase antibody has been validated for the following applications: western blot, ELISA, immunohistochemistry.
Type: Primary
Antigen: GOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Fungi