Anti-FoxJ1/HFH4 Rabbit Polyclonal Antibody
NOVUNBP1-87928
- Antibody type:Primary
- Antigen symbol:FoxJ1/HFH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Gene ID:2302
- Antigen synonyms:HFH-4forkhead box protein J1|FKHL13forkhead transcription factor HFH-4|forkhead-like 13|Hepatocyte nuclear factor 3 forkhead homolog 4|Forkhead-related protein FKHL13|forkhead box J1|HFH4fork head homologue 4|MGC35202
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:PREKDEPGKGGFWRIDPQYAERLLSGAFKKRRLPPVHIHPAFARQAAQEPSAVPRAGPLTVNTEAQQLLREFEEATGEAGWGAGEGRLGHKRKQPLPKRVAKVPRPPSTLLPTPEEQGELEPLKG
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
The FoxJ1 / HFH4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FoxJ1 / HFH4. This antibody reacts with human. The FoxJ1 / HFH4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FoxJ1/HFH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human