Cell detachment solution, Corning® Accutase®
Catalog # 734-2765
Supplier: Corning
Specifications
- Description:Accutase cell detachment solution
- Pk:100 mL
Specifications
About this item
Accutase® is an effective cell detachment solution for the routine detachment of cells from standard tissue culture plasticware as well as enhanced surface and advanced surface coated cultureware
- Efficient, with gentle detachment of adherent cells within minutes
- An alternative to Trypsin
- Able to eliminate neutralization or wash steps
- Convenient and cost effective
Accutase® is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting. It contains proprietary marine-origin enzymes with proteolytic and collagenolytic activity for the detachment of primary and stem cell lines and tissues. Accutase® provides a reduced contamination risk, because it is free of mammalian or bacteria-derived products.