Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium chloride solutions
Magnesium chloride solution 1 M
Magnesium chloride solution 1 M
Magnesium chloride solution 1 M
R004
:
G-Biosciences
:
Magnesium chloride solution 1 M
R004
:
G-Biosciences
:
Pk:
100 mL
Seq:
2440
:
:
203.3 g/mol
:
Ambient
:
7791-18-6
:
MgCl₂·6H₂O
:
MFCD00149781