Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant keratin-8
Human recombinant keratin-8
Human recombinant keratin-8
Catalog # APOSBITP1585-5UG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant keratin-8
Catalog # APOSBITP1585-5UG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
5 µG
Protein/peptide type:
Recombinant
Species:
Human
Protein/peptide name:
Keratin-8
Specifications
About this item
Product Details
Product Details