Order Entry
Slovenia
ContactUsLinkComponent
Human recombinant Cathepsin-L
Human recombinant Cathepsin-L
Catalog # APOSBITP1532-5UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant Cathepsin-L
Catalog # APOSBITP1532-5UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    03
  • Protein synonyms:
    CTSL
  • Protein/peptide name:
    Cathepsin-L

Specifications

About this item