0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Proteins and Peptides
Mouse recombinant FGF-b
Mouse recombinant FGF-b
Mouse recombinant FGF-b
Catalog # APOSBITP1050-10UG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Mouse recombinant FGF-b
Catalog # APOSBITP1050-10UG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
10 µG
Protein/peptide type:
Recombinant
Species:
Mouse
Protein synonyms:
Fibroblast Growth Factor-basic
Protein/peptide name:
FGF-b
Specifications
About this item
Product Details
Product Details