NANOCOLOR® Photometric Tube Test Kit for pH
MANA91872
:
- Chemical method:Phenol red
- Pk:1 KIT
- Description:NANOCOLOR® pH 6,5-8,2, test no. 0-72
- Dimensions:160×110×128 mm (including packaging)
- Model:MANA91872
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:153 g (including packaging)
- For unit:UV/VIS II;VIS II;PF-12Plus;500 D;PF-3 Drinking Water;PF-3 Pool;Advance
- Tests per kit:100
- Range:6,5 - 8,2
A pre-dosed NANOCOLOR® tube test for photometric determination of pH in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with reagents for 100 tests.