
Poly(caprolactone)
Catalog # APOSBIBE1005-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.1400 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:24980-41-4
Formula:(C₆H₁₀O₂)n
MDL Number:MFCD00084404