Order Entry
Slovenia
ContactUsLinkComponent
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
Catalog # APOSPC905485-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
Catalog # APOSPC905485-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.11 g/mol
Storage Temperature:Ambient
CAS Number:119083-00-0
Formula:C₆H₅F₃N₂O₂
MDL Number:MFCD04110182