Order Entry
Slovenia
ContactUsLinkComponent
3-Fluoro-6-methyl-2-nitroaniline 96%
3-Fluoro-6-methyl-2-nitroaniline 96%
Catalog # APOSPC902766-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Fluoro-6-methyl-2-nitroaniline 96%
Catalog # APOSPC902766-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:1393442-58-4
Formula:C₇H₇FN₂O₂
MDL Number:MFCD22383663