
4-(4-Fluorobenzoyl)-1H-pyrrole-2-carboxylic acid
Catalog # APOSPC8782-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.2 g/mol
Melting Pt:223…225 °C
:GHS07
Storage Temperature:Ambient
CAS Number:924868-85-9
Formula:C₁₂H₈FNO₃
MDL Number:MFCD08443967