
Ethyl-2,2,3,3,3-pentafluoropropyl carbonate 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:277332-96-4
:
Formula:C₆H₇F₅O₃
MDL Number:MFCD08688581
:222.11 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together