4-(Trifluoromethoxy)benzoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36823-88-8
:
Formula:C₈H₄ClF₃O₂
MDL Number:MFCD00052329
Boiling Pt:55…56 °C/2mmHg
Density:1.425 g/cm³ (25 °C)
Flash Pt:32 °C
UN:2920
ADR:8+3,II
:224.57 g/mol
:GHS02
:GHS05
Storage Temperature:Ambient
Frequently Bought Together