2-Bromo-5-fluoro-3-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38186-85-5
:
Formula:C₆H₅BrFN
MDL Number:MFCD09835355
:190.01 g/mol
Density:1.598 g/cm³ (25 °C)
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together