

3-Fluoro-4-methoxyphenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:452-11-9
:
Formula:C₇H₇FO₂
MDL Number:MFCD09263471
:142.1277 g/mol
Melting Pt:41…41.5 °C
UN:1759
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together