

5-Bromo-3-fluoropicolinonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886373-28-0
:
Formula:C₆H₂BrFN₂
MDL Number:MFCD07375030
:200.9959 g/mol
Melting Pt:95…100 °C
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Refrigerator
Frequently Bought Together