
5-Bromo-4-chloro-6-(trifluoromethyl)pyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:425392-76-3
:
Formula:C₅HBrClF₃N₂
MDL Number:MFCD09909762
:261.43 g/mol
Boiling Pt:44…45 °C/5mmHg
Melting Pt:–2…0 °C
Density:1.92 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together