
1-Bromo-2,5-bis(trifluoromethyl)benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7617-93-8
:
Formula:C₈H₃BrF₆
MDL Number:MFCD03094336
:293.01 g/mol
Boiling Pt:146…147 °C
Density:1.691 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together