
Pentafluorophenol 99%
:771-61-9
:
:C₆HF₅O
:MFCD00002156
:184.07 g/mol
:143 °C
:34…36 °C
:1.757 g/cm³ (25 °C)
:72 °C
:212-235-8
:GHS07
:Ambient
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Snap Cap Vials
: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sodium triacetoxyborohydride 97%
: Thermo Fisher Scientific
Centrifugal filtration units, Vivaspin® 500
: Sartorius
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
: Apollo Scientific