4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1204296-05-8
:
Formula:C₉H₁₃F₃O₂
MDL Number:MFCD14525503
:210.2 g/mol
Storage Temperature:Ambient
Frequently Bought Together