
4-Fluoro-3-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113421-98-0
:
Formula:C₇H₅F₄NO
MDL Number:MFCD09832246
:195.12 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
Frequently Bought Together