
2-Fluoro-6-iodobenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:111771-08-5
:
Formula:C₇H₄FIO₂
MDL Number:MFCD00042289
:266.01 g/mol
Melting Pt:118…126 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together