
1-(2-Fluoro-5-nitrophenyl)-2,5-dimethylpyrrole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:465514-85-6
:
Formula:C₁₂H₁₁FN₂O₂
MDL Number:MFCD02180649
:234.23 g/mol
Melting Pt:56…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together