Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Bis(2,2,2-trifluoroethyl)ether

Bis(2,2,2-trifluoroethyl)ether

Flammable
Irritant
Supplier:   Apollo Scientific
Bis(2,2,2-trifluoroethyl)ether
Supplier:  Apollo Scientific
Certificates
Bis(2,2,2-trifluoroethyl)ether

2 Options Available Below

Product Details & Documents
Synonyms:  2,2,2-Trifluoroethyl Ether
  • CAS Number:333-36-8
  • Formula:C₄H₄F₆O
  • MDL Number:MFCD00010604
  • Molecular Weight:182.07 g/mol
  • Boiling Pt:62…63 °C
  • Density:1.404 g/cm³ (25 °C)
  • Flash Pt:1 °C
  • Storage Temperature:Ambient
  • UN:3271
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 213,00 €
    213,00 €
    1 * 5 g
       213,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 501,00 €
    501,00 €
    1 * 25 g
       501,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.