2-Fluor-5-methoxy-3-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373921-05-1
:
Formula:C₉H₇BrF₄O
MDL Number:MFCD22201038
:287.05 g/mol
Storage Temperature:Ambient
Frequently Bought Together