
4-Fluoro-N-methoxy-N-methylbenzamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:116332-54-8
:
Formula:C₉H₁₀FNO₂
MDL Number:MFCD02179265
:183.18 g/mol
Melting Pt:31…34 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together