Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Fluorocyclohexanone 96%

2-Fluorocyclohexanone 96%

Supplier:   Apollo Scientific
2-Fluorocyclohexanone 96%
Supplier:  Apollo Scientific
Certificates
2-Fluorocyclohexanone 96%

4 Options Available Below

Product Details & Documents
  • CAS Number:694-82-6
  • Formula:C₆H₉FO
  • MDL Number:MFCD01076476
  • Molecular Weight:116.14 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg

  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 183,00 €
    183,00 €
    1 * 1 g
       183,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 870,00 €
    870,00 €
    1 * 5 g
       870,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.