8-Methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSPC200323-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:244.17 g/mol
Melting Pt:196…197 °C
Storage Temperature:Ambient
CAS Number:874776-53-1
Formula:C₁₀H₇F₃N₂O₂
MDL Number:MFCD03407573