
5-Chloro-2-(trifluoromethyl)benzyl alcohol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261763-21-7
:
Formula:C₈H₆ClF₃O
MDL Number:MFCD01631355
:210.5800 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together