3-Chloro-2-fluorophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2613-22-1
:
Formula:C₆H₄ClFO
MDL Number:MFCD01631572
:146.55 g/mol
Boiling Pt:190 °C
Melting Pt:34…38 °C
EINECS:220-042-5
UN:3261
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together