
2,4-Dimethylpyrrole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:625-82-1
:
Formula:C₆H₉N
MDL Number:MFCD00192088
:95.1424 g/mol
Boiling Pt:165…167 °C
Density:0.924 g/cm³ (25 °C)
Flash Pt:>113 °C
EINECS:210-912-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together