Order Entry
Slovenia
ContactUsLinkComponent
N-Methoxy-N-methylbutyramide 98%
N-Methoxy-N-methylbutyramide 98%
Catalog # APOSOR959865-5G
Supplier:  Apollo Scientific
CAS Number:  
N-Methoxy-N-methylbutyramide 98%
Catalog # APOSOR959865-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :131.18 g/mol
Storage Temperature:Ambient
CAS Number:109480-78-6
Formula:C₆H₁₃NO₂
MDL Number:MFCD14707556