
4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84547-86-4
:
Formula:C₅H₅BrN₂O₂
MDL Number:MFCD00463988
:205.02 g/mol
Melting Pt:200…220 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together