5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34171-37-4
:
Formula:C6H8N2OS
MDL Number:MFCD00079592
:156.2100 g/mol
Storage Temperature:Ambient
Frequently Bought Together