4-Hydroxy-1-cyclohexanecarboxylic acid-δ-lactone 98%
Catalog # APOSOR920857-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.16 g/mol
Boiling Pt:123 °C/11mmHg
Melting Pt:128 °C
Storage Temperature:Ambient
CAS Number:4350-84-9
Formula:C₇H₁₀O₂
MDL Number:MFCD06797111